missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HTR2C Polyclonal Antibody
GREENER_CHOICE

Product Code. 11366363
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366363 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11366363 Supplier Invitrogen™ Supplier No. PA518391

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

Serotonin (5-hydroxytryptamine, 5-HT), a neurotransmitter, elicits a wide array of physiological effects by binding to several receptor subtypes, including the 5-HT2 family of seven-transmembrane-spanning, G-protein-coupled receptors, which activate phospholipase C and D signaling pathways. This gene encodes the 2C subtype of serotonin receptor and its mRNA is subject to multiple RNA editing events, where genomically encoded adenosine residues are converted to inosines. RNA editing is predicted to alter amino acids within the second intracellular loop of the 5-HT2C receptor and generate receptor isoforms that differ in their ability to interact with G proteins and the activation of phospholipase C and D signaling cascades, thus modulating serotonergic neurotransmission in the central nervous system. Studies in humans have reported abnormalities in patterns of 5-HT2C editing in depressed suicide victims.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR2C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene HTR2C
Gene Accession No. P28335
Gene Alias 5ht1c; 5-HT1C; 5-HT-1C; 5HT-1C; 5-HT1c receptor; 5-HT2C; 5-HT-2C; 5HT2C Receptor; 5-HT2C receptor; 5-HT2cR; 5HTR2C; 5-HTR2C; 5-hydroxytryptamine (serotonin) receptor 2C; 5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled; 5-hydroxytryptamine receptor 1C; 5-hydroxytryptamine receptor 2C; HGNC:5295; HTR1C; Htr2c; RP11-810O3.1; RP23-15B7.1; serotonin 1c receptor; serotonin 5-HT-1C receptor; serotonin 5-HT-2C receptor; serotonin receptor 2C; SR1
Gene Symbols HTR2C
Host Species Goat
Immunogen Synthetic peptide sequence (QVENLELPVN) corresponding to the internal amino acids of HTR2C (aa 437-447).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3358
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.