missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTR3B Monoclonal Antibody (2H9H5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531772
This item is not returnable.
View return policy
Description
MA5-31772 has been tested in indirect ELISA.
The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit B of the type 3 receptor for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude.
Specifications
| HTR3B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O95264 | |
| HTR3B | |
| Purified recombinant fragment of human HTR3B (AA: extra 23-150) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 2H9H5 | |
| Unconjugated | |
| HTR3B | |
| 5-HT3 receptor subunit B; 5-HT3B; 5-HT-3B; 5-HT3-B; 5-HT3B receptor; 5-hydroxytryptamine (serotonin) receptor 3B; 5-hydroxytryptamine (serotonin) receptor 3B, ionotropic; 5-hydroxytryptamine 3 receptor B subunit; 5-hydroxytryptamine receptor 3B; HTR3B; Serotonin receptor 3B; serotonin-gated ion channel subunit | |
| Mouse | |
| Protein G | |
| RUO | |
| 9177 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction