missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTR3D Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104300
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HTR3D.
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor is a ligand-gated ion channel, which when activated causes fast, depolarizing responses. It is a cation-specific, but otherwise relatively nonselective, ion channel.
Specifications
| HTR3D | |
| Polyclonal | |
| Unconjugated | |
| HTR3D | |
| 5HT3D; 5-HT3-D; 5-hydroxytryptamine (serotonin) receptor 3 family member D; 5-hydroxytryptamine (serotonin) receptor 3D, ionotropic; 5-hydroxytryptamine receptor 3 subunit D; 5-hydroxytryptamine receptor 3D; HTR3D; serotonin 5-HT-3D receptor; serotonin receptor 3D | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 200909 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q70Z44 | |
| HTR3D | |
| A synthesized peptide derived from human HTR3D(Accession Q70Z44), corresponding to amino acid residues A144-E194. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction