missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTR4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572972
This item is not returnable.
View return policy
Description
HTR4 Polyclonal Antibody for Western Blot, ICC/IF
This gene is a member of the family of serotonin receptors, which are G protein coupled receptors that stimulate cAMP production in response to serotonin (5-hydroxytryptamine). The gene product is a glycosylated transmembrane protein that functions in both the peripheral and central nervous system to modulate the release of various neurotransmitters. Multiple transcript variants encoding proteins with distinct C-terminal sequences have been described, but the full-length nature of some transcript variants has not been determined.
Specifications
| HTR4 | |
| Polyclonal | |
| Unconjugated | |
| HTR4 | |
| 5 hydroxytryptamine ( serotonin) receptor 4; 5 hydroxytryptamine (serotonin) receptor 4; 5-HT>4L>; 5HT4; 5-HT4; 5-HT-4; 5HT4 receptor; 5-HT4 receptor; 5-HT4 receptor (two splicing variants L and S); 5-HT4R; 5HTR4; 5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled; 5-hydroxytryptamine receptor 4; cardiac 5-HT4 receptor; Htr4; Serotonin receptor 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3360 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.09 mg/mL | |
| PBS with 30% glycerol and 0.05% sodium azide | |
| Q13639 | |
| HTR4 | |
| A synthetic peptide made to a C-terminal portion of the human 5HT4 Receptor protein (between residues 350-388). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction