missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTR5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102046
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total 5HT5A receptor.
The neurotransmitter serotonin (5-hydroxytryptamine, 5-HT) has been implicated in a wide range of psychiatric conditions and also has vasoconstrictive and vasodilatory effects. The gene described in this record is a member of 5-hydroxytryptamine (serotonin) receptor family and encodes a multi-pass membrane protein that functions as a receptor for 5-hydroxytryptamine and couples to G-proteins. This protein has been shown to function in part through the regulation of intracellular Ca2+ mobilization.
Specifications
| HTR5A | |
| Polyclonal | |
| Unconjugated | |
| HTR5A | |
| 5-HT-5; 5HT5A; 5-HT5A; 5-HT-5A; 5-HT5a receptor; 5-hydroxytryptamine (serotonin) receptor 5A; 5-hydroxytryptamine (serotonin) receptor 5A, G protein-coupled; 5-hydroxytryptamine receptor 5A; 5-hydroxytryptamine5a; Htr5; Htr5a; MR22; REC17; Serotonin receptor 5A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3361 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P47898 | |
| HTR5A | |
| A synthesized peptide derived from human HTR5A(Accession P47898), corresponding to amino acid residues W221-E271. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction