missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HTR6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15889853
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15889853 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15889853 Supplier Invitrogen™ Supplier No. PA571448

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HTR6 Polyclonal Antibody for Western Blot

This is one of the several different receptors for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that stimulate adenylate cyclase. It has a high affinity for tricyclic psychotropic drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HTR6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HTR6
Gene Accession No. P50406
Gene Alias 5-HT6; 5-HT-6; 5-HT6 receptor; 5-HT6R; 5-hydroxytryptamine (serotonin) receptor 6; 5-hydroxytryptamine (serotonin) receptor 6, G protein-coupled; 5-hydroxytryptamine receptor 6; Htr6; Serotonin receptor 6; ST-B17
Gene Symbols HTR6
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human HTR6.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3362
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.