missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HTRA1/PRSS11 Antibody (FHTR2163), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28037-50ug
This item is not returnable.
View return policy
Description
HTRA1/PRSS11 Monoclonal antibody specifically detects HTRA1/PRSS11 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| HTRA1/PRSS11 | |
| Monoclonal | |
| Unconjugated | |
| Q92743 | |
| Human | |
| Protein A purified | |
| RUO | |
| 5654 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| FHTR2163 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| ARMD7, EC 3.4.21, EC 3.4.21.108, High-temperature requirement A serine peptidase 1, HtrA serine peptidase 1, L56, ORF480, Serine protease 11, serine, 11 (IGF binding) | |
| HTRA1 | |
| 50 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction