missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HTRA1/PRSS11 Antibody (SR4329), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27848-50ul
This item is not returnable.
View return policy
Description
HTRA1/PRSS11 Monoclonal antibody specifically detects HTRA1/PRSS11 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| HTRA1/PRSS11 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5654 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR4329 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| ARMD7, EC 3.4.21, EC 3.4.21.108, High-temperature requirement A serine peptidase 1, HtrA serine peptidase 1, L56, ORF480, Serine protease 11, serine, 11 (IGF binding) | |
| A synthesized peptide derived from human HTRA1/PRSS11 (Uniprot #: Q92743) | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction