missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTRA2/Omi Antibody (196C429), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56558AF750
This item is not returnable.
View return policy
Description
HTRA2/Omi Monoclonal specifically detects HTRA2/Omi in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HTRA2/Omi | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HTRA2 | |
| This monoclonal antibody was developed by immunizing mice with a synthetic peptide containing amino acids 335-350 of human HtrA2. | |
| Primary | |
| This antibody does not cross-react with mouse HtrA2. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 196C429 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI | |
| Mouse | |
| 0.1 mL | |
| 27429 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction