missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTRA2/Omi Antibody (1B3), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04325G
This item is not returnable.
View return policy
Description
HTRA2/Omi Monoclonal specifically detects HTRA2/Omi in Human samples. It is validated for Western Blot, ELISA.
Specifications
| HTRA2/Omi | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HTRA2 | |
| Recombinant human HtrA2/Omi (134-458 aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, Cancer, Mitochondrial Mediated Pathway | |
| 27429 | |
| Human, Mouse, Rat (Negative) | |
| Purified |
| Western Blot, ELISA | |
| 1B3 | |
| Western Blot, ELISA | |
| High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human HtrA2/Omi mAb, clone 1B3, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human HtrA2/Omi protein. Clone 1B3 is specific against human and mouse, but not rat. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction