Learn More
HTRA2 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15775-1-AP-20UL
 View more versions of this product
Product Code. 16855104
  • € 145.00 / 20µL
Description
The inhibitor of apoptosis (IAP) family of proteins regulates programmed cell death triggered by various stimuli. All IAPs have at least one baculovirus IAP repeat (BIR) motif that is essential for their anti-apoptotic activity (1-3). Recently, a serine protease has been isolated, which is released from the mitochondria upon induction of apoptosis by apoptotic stimuli. This protein called Omi/HtrA2 is a mitochondrial protein that binds to IAP. Like Smac/DIABLO, the mature Omi protein contains a conserved IAP-binding motif (AVPS) at its N terminus. The deregulated expression of Omi in the cytoplasm of mammalian cells induces apoptosis indicating that Omi could participate in the mitochondrial apoptotic pathway.
Specifications
Specifications
HTRA2
Polyclonal
Unconjugated
HTRA2
HtrA serine peptidase 2, HTRA2, OMI, PARK13, PRSS25, Serine protease 25, Serine proteinase OMI
Rabbit
Antigen Affinity Chromatography
RUO
27429, 297376, 64704
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43464, Q9JIY5
Htra2
HTRA2 Fusion Protein Ag8455
20 μL
Primary
Mouse, Human, Non-human Primate, Rat