missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Bovine Albumin ELISA Kit (Colorimetric)

Product Code. 18618956 Shop All Bio Techne Products
Click to view available options
Quantity:
96 wells
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18618956

Brand: Novus Biologicals™ NBP312180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.69 to 500 ng/mL
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Colostrum, Milk, Cell Culture Supernatant
Target Species Bovine
For Use With (Application) ELISA
Gene ID (Entrez) 213
Gene Symbol ALB
Quantity 96 wells
Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Storage Requirements Store at 4°C.
Synonym albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Target Bovine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.