missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human A33 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
€ 207.00 - € 587.00
Specifications
| Antigen | A33/GPA33 |
|---|---|
| Clone | 2963D |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.10 ug/10^6 cells |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18161971
|
R&D Systems
MAB11365-100 |
100 μg |
€ 587.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18131981
|
R&D Systems
MAB11365-SP |
25 μg |
€ 207.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A33/GPA33 Monoclonal antibody specifically detects A33/GPA33 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| A33/GPA33 | |
| Western Blot 1 ug/mL, Flow Cytometry 0.10 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), GPA33, MGC129986, MGC129987, transmembrane glycoprotein A33 | |
| E. coli-derived recombinant human A33, Ile22-Val235, Accession # Q99795 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2963D | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q99795 | |
| 10223 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title