missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Active MMP-1 Fluorokine E Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The Fluorokine E Human Active MMP-1 kit combines the specificity of a monoclonal antibody that captures both pro and active MMP-1 forms and the sensitivity of fluorescence. The kit is designed to measure the levels of both endogenous active MMP-1 in cell culture supernates, serum, and plasma and the MMP-1 in these samples that can be activated by APMA in the assay procedure. The measured MMP-1 activity may reflect the balance of MMP-1 and its inhibitor TIMPs. Therefore, it is recommended that the levels of TIMPs be determined in order to interpret the results properly. For example, TIMP-1 inhibits the activity of MMP-1 through binding of the N-terminal domain of the TIMP-1 to the active site of MMP-1 (8). The Quantikine Human TIMP-1 Immunoassay (R&D Systems Catalog # DTM100) can be used to measure TIMP-1 concentrations in these samples.
Specifications
Specifications
| Assay Range | 0.39 to 25 pg/mL |
| Assay Sensitivity | 0.15 pg/mL |
| Gene ID (Entrez) | 4312 |
| Gene Symbol | MMP1 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP1, MMP-1 |
| Target | MMP-1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction