missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM8 aa 498-653 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC10311U-100UG
This item is not returnable.
View return policy
Description
ADAM8 Monoclonal specifically detects ADAM8 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| ADAM8 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P78325 | |
| ADAM8 | |
| Mouse myeloma cell line NS0-derived recombinant human ADAM8 ectodomain Asp158-Ser653 Accession # P78325 | |
| RUO | |
| 101 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 143338 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2, MS2disintegrin and metalloproteinase domain-containing protein 8 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ADAM8 in direct ELISAs and Western blots. Detects aa 498-653 of rhADAM8, which corresponds to the cysteine-rich and EGF-like domains. In direct ELISAs, no cross-reactivity with recombinant human (rh) ADAM9, rhADAM10, rhADAM15, rhBACE, or rhTACE is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction