missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM8 Ectodomain Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF1031
This item is not returnable.
View return policy
Description
ADAM8 Polyclonal specifically detects ADAM8 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.
Specifications
| ADAM8 | |
| Polyclonal | |
| Unconjugated | |
| P78325 | |
| ADAM8 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ADAM8 Ectodomain Glu158-Ser653 Accession # P78325 | |
| 100 μg | |
| Primary | |
| Detects human ADAM8 Ectodomain in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse ADAM8 is observed and less than 1% cross-reactivity with recombinant human (rh) ADAM12, rhADAM19, and rhADAM33 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| a disintegrin and metalloproteinase domain 8, ADAM 8, ADAM metallopeptidase domain 8, CD156a, CD156a antigen, CD156human leukocyte differentiation antigen, Cell surface antigen MS2, EC 3.4.24, EC 3.4.24.-, MGC134985, MS2, MS2disintegrin and metalloproteinase domain-containing protein 8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 101 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction