missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAMTS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2197-SP
This item is not returnable.
View return policy
Description
ADAMTS1 Monoclonal specifically detects ADAMTS1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ADAMTS1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q9VHI8 | |
| ADAMTS1 | |
| Mouse myeloma cell line NS0-derived recombinant human ADAMTS1 Phe253-Cys724 (predicted) Accession # Q9VHI8 | |
| 25 μg | |
| Primary | |
| Detects human ADAMTS1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human ADAMTS-L1.2 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 268104 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1, ADAM metallopeptidase with thrombospondin type 1 motif, 1, ADAM-TS 1, ADAM-TS1, ADAMTS-1, C3-C5, METH1, METH-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9510 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction