missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Adenylate Cyclase 9 (aa 299-393) Control Fragment Recombinant Protein

Product Code. 30182378
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182378

Brand: Invitrogen™ RP98698

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83713 (PA5-83713. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. It is regulated by a family of G protein-coupled receptors, protein kinases, and calcium. The type 9 adenylyl cyclase is a widely distributed adenylyl cyclase, and it is stimulated by beta-adrenergic receptor activation but is insensitive to forskolin, calcium, and somatostatin.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60503
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 115
Name Human Adenylate Cyclase 9 (aa 299-393) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AC9; ACIX; ACtp10; Adcy9; adenylate cyclase 9; adenylate cyclase type 9; Adenylate cyclase type IX; adenylyl cyclase 9; Adenylyl cyclase type 10; ATP pyrophosphate-lyase 9; AW125421; D16Wsu65e; KIAA0520; mKIAA0520; type IX ATP pyrophosphate-lyase
Common Name Adenylate Cyclase 9
Gene Symbol ADCY9
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FVMSQVRSRSTFLKVGQSIMHGKDLEVEKALKERMIHSVMPRIIADDLMKQGDEESENSVKRHATSSPKNRKKKSSIQKAPIAFRPFKMQQIEEV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.