missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Adiponectin/Acrp30 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1065
This item is not returnable.
View return policy
Description
Adiponectin/Acrp30 Polyclonal specifically detects Adiponectin/Acrp30 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| Adiponectin/Acrp30 | |
| Polyclonal | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| ACDC, Acrp30, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, ApM1, APM1APM-1, C1q and collagen domain-containing protein, GBP28, GBP28apM1, Gelatin-binding protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9370 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| Unconjugated | |
| Q15848 | |
| ADIPOQ | |
| Mouse myeloma cell line NS0-derived recombinant human Adiponectin/Acrp30 Glu19-Asn244 Accession # Q15848 | |
| 100 μg | |
| Primary | |
| Detects human Adiponectin/Acrp30 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse Adiponectin is observed and less than 5% cross-reactivity with recombinant rat Adiponectin is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction