missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Afamin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8065
This item is not returnable.
View return policy
Description
Afamin Monoclonal specifically detects Afamin in Human samples. It is validated for Western Blot.
Specifications
| Afamin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P43652 | |
| AFM | |
| HEK293 human embryonic kidney cell line transfected with human Afamin Leu22-Asn599 Accession # P43652 | |
| 100 μg | |
| Primary | |
| Detects human Afamin in ELISA and Western blot. | |
| Human | |
| Purified |
| Western Blot | |
| 883113 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| afamin, AFM, ALB2, ALB2alpha-Alb, ALBA, ALBAalpha-albumin, ALF, Alpha-Alb, Alpha-albumin, MGC125338, MGC125339 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 173 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction