missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Aggrecan (aa 2133-2218) Control Fragment Recombinant Protein

Product Code. 30208464
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208464

Brand: Invitrogen™ RP105949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (61%), Rat (61%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Aggrecan also known as cartilage-specific proteoglycan core protein (CSPCP) or chondroitin sulfate proteoglycan 1 is a protein that is encoded by the ACAN gene. The encoded protein is an integral part of the extracellular matrix in cartilaginous tissue and it withstands compression in cartilage. The human form of the protein is 2316 amino acids long (approximately 250 kDa) and can be expressed in multiple isoforms due to alternative splicing. Aggrecan exhibits a bottlebrush structure, in which chondroitin sulfate and keratan sulfate chains are attached to an extended protein core. The synthesis and degradation of aggrecan are being investigated for their roles in cartilage deterioration during joint injury, disease, and aging. It contains three interglobular domains (IGDs), G1, G2, and G3 that are involved in aggregation, hyaluronan binding, cell adhesion, and chondrocyte apoptosis. Aggrecan fragments from articular cartilage are released into the synovial fluid at all stages of human osteoarthritis as a result of cleavage by aggrecanase and MMP activity. Antibodies BC-3 and BC-4 can be used to detect these neo epitopes of Aggrecanase (ARGSVXXX) and MMP (XXXVDIPEN), respectively.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P16112
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 176
Name Human Aggrecan (aa 2133-2218) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ACAN; Agc; Agc1; AGCAN; aggrecan; aggrecan 1; aggrecan 1 (chondroitin sulfate proteoglycan 1, large aggregating proteoglycan, antigen identified by monoclonal antibody A0122); aggrecan core protein; Aggrecan core protein 2; aggrecan CS2 domain; aggrecan epidermal growth factor-like domain-1; aggrecan G2 domain; aggrecan interglobular domain; aggrecan keratan sulfate domain; aggrecan, structural proteoglycan of cartilage; articular cartilage aggrecan precursor; b2b183Clo; cartilage aggregating proteoglycan; cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; chondroitin sulfate proteoglycan core protein 1; cmd; CSPCP; CSPG1; CSPGCP; high density cartilage proteoglycan; large aggregating cartilage proteoglycan core protein; large aggregating proteoglycan; large aggregating proteoglycan, antigen; MSK16; SEDK
Common Name Aggrecan
Gene Symbol ACAN
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TSAFHEANLERSSGLGVSGSTLTFQEGEASAAPEVSGESTTTSDVGTEAPGLPSATPTASGDRTEISGDLSGHTSQLGVVISTSIP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.