Learn More
Human Akt Pan Specific Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  R&D Systems AF2055-SP
Product Code. 18769127
  • ONLINE EXCLUSIVE PRICE
    € 277.00
    € 261.45 / 25µg
    Save € 15.55
    5% Off
Description
AKT Polyclonal specifically detects AKT in Human samples. It is validated for Western Blot, Simple Western.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AKT
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB alpha, PKB beta, PKB gamma, PKB-ALPHA, PRKBA, RAC, RAC-alpha, RAC-beta, RAC-gamma
Rabbit
Affinity Purified
RUO
207
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 0.1 ug/mL, Simple Western 20 ug/mL
P31749
AKT1
Akt synthetic peptide IRCLQWTTVIERTFHV Accession # P31749
25 μg
Primary
Detects human Akt1, Akt2, and Akt3 in Western blots.