missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Akt1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB17751
This item is not returnable.
View return policy
Description
AKT1 Monoclonal specifically detects AKT1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| AKT1 | |
| Monoclonal | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| P31749 | |
| AKT1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Akt1 Ser2-Ala480 Accession # P31749 | |
| 100 μg | |
| Primary | |
| Detects human Akt1 in direct ELISAs. Reactivity with Akt1 from other species has not been determined. In direct ELISAs, no cross-reactivity with recombinant human (rh) Akt2 or rhAkt3 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 302407 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 207 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction