missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Akt1 (E17K Mutation) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6815-SP
This item is not returnable.
View return policy
Description
AKT1 Monoclonal specifically detects AKT1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| AKT1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P31749 | |
| AKT1 | |
| Synthetic peptide containing human Akt1 Ala5-Lys30 with Glu17Lys mutation Accession # P31749 | |
| 25 μg | |
| Primary | |
| Detects human AKT1 with Glu17Lys mutation in Western blots. In Western blots, no cross-reactivity with 293-EBNA human EBV-expressing embryonic kidney cell line transfectant expressing wild type AKT1 is detected. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 710022 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 207 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction