missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Alkaline Phosphatase/ALPP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB59051-SP
This item is not returnable.
View return policy
Description
Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Alkaline Phosphatase/ALPP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P05187 | |
| ALPP | |
| Mouse myeloma cell line NS0-derived recombinant human Alkaline Phosphatase/ALPP Ile23-Asp506 Accession # P05187 | |
| 25 μg | |
| Primary | |
| Detects human Alkaline Phosphatase/ALPP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Alkaline Phosphatase/ALPI is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 696128 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Alkaline Phosphatase, Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme, SEAP | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 250 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction