missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human alpha 1-Microglobulin (NP_001624) VersaClone cDNA

Product Code. 18734768
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18734768 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18734768 Supplier R&D Systems™ Supplier No. RDC2401
  • ONLINE EXCLUSIVE PRICE
    € 341.00
    € 322.35 / 10µg
    Save € 18.65
    5% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ Human alpha 1-Microglobulin (NP_001624) VersaClone cDNA is a glycoprotein secreted in plasma.

The Human alpha 1-Microglobulin (NP_001624) VersaClone cDNA clone contains the complete ORF for alpha 1-Microglobulin, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The alpha 1-Microglobulin insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Specifications

Gene hAMBP
Accession Number NP_001624
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 1072bp
pH 8.5
Gene ID (Entrez) 259
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.