missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human alpha-1a Adrenergic Receptor (aa 2-27) Control Fragment Recombinant Protein

Product Code. 30203175
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203175

Brand: Invitrogen™ RP109585

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Adrenergic receptors (ARs) are members of the 7-transmembrane domain G-protein-coupled receptor superfamily that bind the endogenous catecholamines epinephrine and norepinephrine. Pharmacological, structural, and molecular cloning data indicate significant heterogeneity within this receptor family. Nine receptor subtypes have been identified thus far including three alpha-1 AR subtypes (1A/D, 1B, and 1C), three alpha-2 ARs (2A, 2B, and 2C), and three beta AR subtypes (1, 2, and 3). ARs participate in either the onset or maintenance of several disease states including hypertension, cardiac dysfunction (congestive heart failure, ischemia, arrhythmias), diabetes, glaucoma, depression, and impotence. A1AR subtypes are found in numerous tissues and have been shown to be involved in the regulation of blood pressure due to changes in vascular tone and cardiac output. Effects on uterine contraction, hepatic glucose metabolism, heat shock protein 70 (HSP70), proto-oncogene expression, and mitogenesis have been linked to A1AR activation. Norepinephrine can increase nitric oxide synthase (NOS) levels in the hypothalamus by activating alpha-1 AR which may indirectly suppress the release of luteinizing hormone (LH), the androgens, estrogen, and progesterone.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35348
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 148
Name Human alpha-1 A Adrenergic Receptor (aa 2-27) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ADA1; adra1a; adra1aa; Adra1c; ADRA1L1; adrenergic alpha 1 c receptor; adrenergic receptor alpha 1 c subtype; adrenergic receptor, alpha 1 A; adrenergic receptor, alpha 1 c; adrenergic, alpha-1 A-, receptor; adrenergic, alpha-1 Aa, receptor; adrenoceptor alpha 1 A; adrenoceptor alpha 1 Aa; alpha 1 A-adrenoceptor; alpha 1 A-adrenoreceptor; alpha 1 C-adrenergic receptor; alpha-1 A adrenergic receptor; Alpha-1 A adrenoceptor; alpha-1 A adrenoreceptor; ALPHA1AAR; Alpha-1 C adrenergic receptor; Alpha1C-AR; Alpha-adrenergic receptor 1 c; fc18h12; G protein coupled receptor; wu:fc18h12
Common Name alpha-1 A Adrenergic Receptor
Gene Symbol ADRA1A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VFLSGNASDSSNCTQPPAPVNISKAI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.