missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha 1B-Glycoprotein Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7757-SP
This item is not returnable.
View return policy
Description
alpha 1B-Glycoprotein Monoclonal specifically detects alpha 1B-Glycoprotein in Human samples. It is validated for Western Blot.
Specifications
| alpha 1B-Glycoprotein | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P04217 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 875431 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| A1BG | |
| HEK293 human embryonic kidney cell line transfected with human alpha 1B-Glycoprotein Ala22-Ser495 Accession # P04217 | |
| 25 μg | |
| Primary | |
| Detects human alpha 1B-Glycoprotein in ELISAs and Western blot. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction