missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha 2-Macroglobulin Antibody, R&D Systems™

Mouse Monoclonal Antibody
€ 207.00 - € 578.00
Specifications
| Antigen | alpha 2-Macroglobulin |
|---|---|
| Clone | 257303 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18723407
![]() |
R&D Systems
MAB19381-SP |
25 μg |
€ 207.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18710117
|
R&D Systems
MAB19381-100 |
100 μg |
€ 578.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha 2-Macroglobulin Monoclonal antibody specifically detects alpha 2-Macroglobulin in Human samples. It is validated for ELISASpecifications
| alpha 2-Macroglobulin | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 257303 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| A2M, alpha 2Macroglobulin, Alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5, CPAMD5DKFZp779B086, FWP007, S863-7 | |
| Human plasma-derived alpha 2-Macroglobulin | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title