Learn More
Human alpha-2A Adrenergic R/ADRA2A Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB10129U-100UG
This item is not returnable.
View return policy
Description
alpha-2A Adrenergic R/ADRA2A Monoclonal specifically detects alpha-2A Adrenergic R/ADRA2A in Human samples. It is validated for Flow Cytometry.
Specifications
| alpha-2A Adrenergic R/ADRA2A | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human alpha-2A Adrenergic R/ADRA2A in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 1006614 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| a-2A AdrenergicR, ADRA2, ADRA2A, ADRA2Ralpha-2-adrenergic receptor, platelet type, ADRARZNF32, adrenergic, alpha-2A-, receptor, Alpha-2 adrenergic receptor subtype C10, alpha-2A adrenergic receptor, alpha2A AdrenergicR, Alpha-2A adrenoceptor, Alpha-2A adrenoreceptor, Alpha-2AAR, ALPHA2AAR, alpha-2AAR subtype C10, ZNF32 | |
| Synthetic peptide containing Human alpha-2A Adrenergic R/ADRA2A | |
| 100 μg | |
| 150.0 | |
| Human | |
| Supernatant |