missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human alpha Adducin (aa 521-605) Control Fragment Recombinant Protein

Product Code. 30202467
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202467

Brand: Invitrogen™ RP102685

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta, gamma). Adducin is a heterodimeric protein that consists of related subunits, which are produced from distinct genes but share a similar structure. Alpha- and beta-adducin include a protease-resistant N-terminal region and a protease-sensitive, hydrophilic C-terminal region. Alpha- and gamma-adducins are ubiquitously expressed. In contrast, beta-adducin is expressed at high levels in brain and hematopoietic tissues. Adducin binds with high affinity to Ca(2+)/calmodulin and is a substrate for protein kinases A and C. Alternative splicing results in multiple variants encoding distinct isoforms; however, not all variants have been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35611
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 118
Name Human alpha Adducin (aa 521-605) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Add1; ADD2; ADDA; ADDB; adducin 1; adducin 1 (alpha); Adducin beta; Adducin1; Adducin-1; Adducin-alpha; Adducin-beta; AI256389; Alpha adducin; alpha-ADD; alpha-adducin; beta Adducin; Beta-adducin; erythrocyte adducin alpha subunit; erythrocyte adducin subunit alpha
Common Name alpha Adducin
Gene Symbol ADD1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SQVLCGVVMDRSLVQGELVTASKAIIEKEYQPHVIVSTTGPNPFTTLTDRELEEYRREVERKQKGSEENLDEAREQKEKSPPDQP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.