Learn More
Human alpha-Fetoprotein/AFP Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB13691
 View more versions of this product
Product Code. 18171621
  • € 542.00 / 500µg
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha-Fetoprotein/AFP
Monoclonal
LYOPH
Western Blot 1 ug/mL
AFP, Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, DSCAM2, FETA, HP, HPAFP
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
174
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Western Blot
214107
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AFP
Human umbilical cord serum-derived alpha-Fetoprotein/AFP
500 μg
Primary
Detects purified human alpha-Fetoprotein/AFP in Western blots.