Learn More
R&D Systems™ Human alpha-Fetoprotein DuoSet ELISA
Product Code. 30021220
Click to view available options
Conjugate:
Biotin
Unit Size:
1 set
Product Code. 30021220
Brand: R&D Systems™ DY1369
Sufficient for at least 15 96-well plates
The Human alpha-Fetoprotein DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human alpha-Fetoprotein/AFP.
TRUSTED_SUSTAINABILITY
Specifications
Conjugate Biotin
Assay Range 78.10 to 5000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 174
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.