Learn More
Invitrogen™ Human Alpha-Galactosidase A/GLA ELISA Kit
Code produit. 17134979
missing translation for 'orderingAttributeHoverText'
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
missing translation for 'unitSize'
Each
Pack of 10
Pack of 96
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 17134979
missing translation for 'mfr': Invitrogen™ EH21RBX5
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
ELISA
Human Alpha-Galactosidase A/GLA quantitates human Alpha-Galactosidase A/GLA in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Alpha-Galactosidase A/GLA.
This gene encodes a homodimeric glycoprotein that hydrolyses the terminal alpha-galactosyl moieties from glycolipids and glycoproteins. This enzyme predominantly hydrolyzes ceramide trihexoside, and it can catalyze the hydrolysis of melibiose into galactose and glucose. A variety of mutations in this gene affect the synthesis, processing, and stability of this enzyme, which causes Fabry disease, a rare lysosomal storage disorder that results from a failure to catabolize alpha-D-galactosyl glycolipid moieties.
TRUSTED_SUSTAINABILITY
Spécification
Accession Number Agalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; melibiase
Assay Range 45 to 10,000 pg/mL
Assay Sensitivity 45 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2717
Gene Symbol GLA
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target alpha Galactosidase
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins