missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Aminopeptidase P2/XPNPEP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2490
This item is not returnable.
View return policy
Description
Aminopeptidase P2/XPNPEP2 Monoclonal specifically detects Aminopeptidase P2/XPNPEP2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Aminopeptidase P2/XPNPEP2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Aminoacylproline aminopeptidase, APP2, EC 3.4.11.9, mAmP, Membrane-bound aminopeptidase P, Membrane-bound AmP, Membrane-bound APP, xaa-Pro aminopeptidase 2, XPNPEP2, X-Pro aminopeptidase 2, X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7512 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunoprecipitation | |
| 391118 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| AAB96394 | |
| XPNPEP2 | |
| Mouse myeloma cell line NS0-derived recombinant human Aminopeptidase P2/XPNPEP2 His22-Ala650 Accession # AAB96394 | |
| 100 μg | |
| Primary | |
| Detects human Aminopeptidase P2/XPNPEP2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse Aminopeptidase P2/XPNPEP2 and no cross-reactivity with recombinant human XPNPEP1 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction