missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Amphiregulin Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AB-262-NA
This item is not returnable.
View return policy
Description
Amphiregulin Polyclonal specifically detects Amphiregulin in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifications
| Amphiregulin | |
| Polyclonal | |
| Unconjugated | |
| P15514 | |
| AREG | |
| E. coli-derived recombinant human Amphiregulin Val107-Lys184 Accession # P15514 | |
| 1 mg | |
| Primary | |
| Detects human Amphiregulin in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) EGF, rhTGF-alpha , and rhHB-EGF is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 5-20 ug/mL | |
| amphiregulin, AR, AREG, AREGB, Colorectum cell-derived growth factor, CRDGF, MGC13647, schwannoma-derived growth factor, SDGF | |
| Goat | |
| Protein A or G purified | |
| RUO | |
| 374 | |
| Reconstitute at 1 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction