missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Angiopoietin-2 Quantikine ELISA Kit
Klik for at se tilgængelige muligheder
Quantity:
1 Plate
6 Plates
Pakningsstørrelse:
1 set
Beskrivelse
The Human Angiopoietin-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Angiopoietin-2 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (50 ul).
Tekniske data
Tekniske data
| Assay Range | 46.90 - 3000pg/mL |
| Assay Sensitivity | 21.3pg/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum |
| Specificity | Natural and Recombinant Human Angiopoietin-2 |
| Sufficient For | Sufficient materials to run ELISAs on at least one 96-well plate |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion