missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-like Protein 1/ANGPTL1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2204
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 1/ANGPTL1 Monoclonal specifically detects Angiopoietin-like Protein 1/ANGPTL1 in Human samples. It is validated for Western Blot.
Specifications
| Angiopoietin-like Protein 1/ANGPTL1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ANG-3, ANG3angioarrestin, Angiopoietin like Protein 1, angiopoietin Y1, angiopoietin-3, angiopoietin-like 1, Angiopoietin-like protein 1, angiopoietin-related protein 1, ANGPT3, ANGPTL1, AngY, AngY1, ARP1, ARP1angiopoietin 3, dJ595C2.2, dJ595C2.2 (angiopoietin Y1), KIAA0351, UNQ162 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9068 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 205812 | |
| Western Blot 1 ug/mL | |
| O95841 | |
| ANGPTL1 | |
| Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like Protein 1/ANGPTL1 Gly24-Asp491 (predicted) Accession # O95841 | |
| 500 μg | |
| Primary | |
| Detects human Angiopoietin-like Protein 1/ANGPTL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Angiopoietin-1, -2, or -4 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction