missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-like Protein 1/ ANGPTL1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2204-SP
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 1/ANGPTL1 Polyclonal specifically detects Angiopoietin-like Protein 1/ANGPTL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Angiopoietin-like Protein 1/ANGPTL1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ANG-3, ANG3angioarrestin, Angiopoietin like Protein 1, angiopoietin Y1, angiopoietin-3, angiopoietin-like 1, Angiopoietin-like protein 1, angiopoietin-related protein 1, ANGPT3, ANGPTL1, AngY, AngY1, ARP1, ARP1angiopoietin 3, dJ595C2.2, dJ595C2.2 (angiopoietin Y1), KIAA0351, UNQ162 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9068 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL | |
| O95841 | |
| ANGPTL1 | |
| Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 1/ANGPTL1 Ala39-Asp491 Accession # O95841 | |
| 25 μg | |
| Primary | |
| Detects human Angiopoietin-like 1/ANGPTL1 in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human (rh) Angiopoietin-4 is observed and less than 5% cross-reactivity with rhAngiopietin-1 and rhAngiopoietin-2 is observed. | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering