missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ANKS6 (aa 726-855) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52550 (PA5-52550. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
ANKS6 is required for renal function.
Specifications
Specifications
| Accession Number | Q68DC2 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 203286 |
| Name | Human ANKS6 (aa 726-855) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2210417J20Rik; Ankrd14; ANKS6; Ankyrin repeat and SAM domain-containing protein 6; ankyrin repeat and sterile alpha motif domain containing 6; ankyrin repeat domain 14; Ankyrin repeat domain-containing protein 14; b2b1801.1 Clo; Gm635; NPHP16; PKDR1; polycystic kidney disease protein 1; SAM domain-containing protein 6; samcystin; SAMD6; sterile alpha motif domain containing 6; sterile alpha motif domain-containing protein 6 |
| Common Name | ANKS6 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction