missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Ankyrin B (aa 3726-3869) Control Fragment Recombinant Protein

Product Code. 30202469
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202469

Brand: Invitrogen™ RP102424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83300 (PA5-83300. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the ankyrin family of proteins that link the integral membrane proteins to the underlying spectrin-actin cytoskeleton. Ankyrins play key roles in activities such as cell motility, activation, proliferation, contact and the maintenance of specialized membrane domains. Most ankyrins are typically composed of three structural domains: an amino-terminal domain containing multiple ankyrin repeats; a central region with a highly conserved spectrin binding domain; and a carboxy-terminal regulatory domain which is the least conserved and subject to variation. The protein encoded by this gene is required for targeting and stability of Na/Ca exchanger 1 in cardiomyocytes. Mutations in this gene cause long QT syndrome 4. Multiple transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q01484
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 287
Name Human Ankyrin B (aa 3726-3869) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI835472; Ank2; ANK-2; Ank3; AnkB; ankyrin 2, brain; ankyrin 2, neuronal; ankyrin 3, epithelial; ankyrin B; ankyrin, brain; ankyrin-2; ankyrin-2, nonerythrocytic; ankyrin-B; AW491075; Brain ankyrin; brank-2; LQT4; Non-erythroid ankyrin
Common Name Ankyrin B
Gene Symbol ANK2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DGVPKTEGDSSATALFPQTHKEQVQQDFSGKMQDLPEESSLEYQQEYFVTTPGTETSETQKAMIVPSSPSKTPEEVSTPAEEEKLYLQTPTSSERGGSPIIQEPEEPSEHREESSPRKTSLVIVESADNQPETCERLDEDAAFE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.