missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Annexin A4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4146-SP
This item is not returnable.
View return policy
Description
Annexin A4 Monoclonal specifically detects Annexin A4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Annexin A4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P09525 | |
| ANXA4 | |
| E. coli-derived recombinant human Annexin A4 Met1-Asp319 Accession # P09525 | |
| 25 μg | |
| Primary | |
| Detects human Annexin A4 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Annexin A1, A2, A3, A5, A6, A8, A9, A10, A11, or 13A is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 694624 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AIV, annexin A4, Annexin IV, annexin IV (placental anticoagulant protein II), annexin-4, ANX4, ANX4Carbohydrate-binding protein p33/p41, ANXA4, Chromobindin-4, chromobindin-4,35-beta calcimedin, DKFZp686H02120, Endonexin I, Lipocortin IV, MGC75105, P32.5, PAP-II, PIG28, Placental anticoagulant protein II, PP4-X, proliferation-inducing gene 28, proliferation-inducing protein 28, Protein II, ZAP36 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 307 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction