missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human APC1 (aa 1112-1201) Control Fragment Recombinant Protein

Product Code. 30194611
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30194611

Brand: Invitrogen™ RP96147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83323 (PA5-83323. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC1, also known as mitotic checkpoint regulator (MCPR), is a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. The individual APC/C components mRNA and protein levels are expressed at approximately the same levels in most tissues and cell lines, suggesting that they perform their functions as part of a complex.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H1A4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 64682
Name Human APC1 (aa 1112-1201) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610021O03Rik; AI047775; AI853536; ANAPC1; anaphase promoting complex subunit 1; anaphase-promoting complex 1 (meiotic checkpoint regulator); anaphase-promoting complex subunit 1; APC1; AW547281; Cyclosome subunit 1; Mcpr; meiotic check point regulator; mitotic checkpoint regulator; Testis-specific gene 24 protein; TSG24
Common Name APC1
Gene Symbol ANAPC1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PPRNTTVDLNSGNIDVPPNMTSWASFHNGVAAGLKIAPASQIDSAWIVYNKPKHAELANEYAGFLMALGLNGHLTKLATLNIHDYLTKGH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.