missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APJ Alexa Fluor 350 Antibody (Clone 72133R), R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8561U-100UG
This item is not returnable.
View return policy
Description
APJ/Apelin receptor Monoclonal specifically detects APJ/Apelin receptor in Human samples. It is validated for Flow Cytometry.
Specifications
| APJ/Apelin receptor | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| AGTRL1, AGTRL1HG11, angiotensin II receptor-like 1, Angiotensin receptor-like 1, apelin receptor, APJ (apelin) receptor, APJ receptor, APJFLJ96609, APJR, APLNR, FLJ90771, G protein-coupled receptor APJ, G-protein coupled receptor APJ, G-protein coupled receptor HG11, HG11, HG11 orphan receptor, MGC45246 | |
| Mouse myeloma cell line NS0-derived recombinant human APJ Met1-Asp380 Accession # P35414 | |
| RUO | |
| 187 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 72133R | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P35414 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human APJ in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction