missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APLP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB49451-SP
This item is not returnable.
View return policy
Description
APLP-2 Monoclonal specifically detects APLP-2 in Human samples. It is validated for Western Blot.
Specifications
| APLP-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q06481 | |
| APLP2 | |
| Mouse myeloma cell line NS0-derived recombinant human APLP-2 Leu30-Ser680 Accession # Q06481 | |
| 25 μg | |
| Primary | |
| Detects human APLP-2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse APLP-2 or recombinant human APLP-1 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 397702 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP2, APLP-2, APPH, APPHCDEI box-binding protein, APPL2, CDEBP, WALPLP2, YWK-II | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 334 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction