missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Apolipoprotein B/ApoB Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB41241-100
This item is not returnable.
View return policy
Description
Apolipoprotein B/ApoB Monoclonal specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
| Apolipoprotein B/ApoB | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Apo B-100, APOB, apoB-100, ApoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100 | |
| E. coli-derived recombinant human Apolipoprotein B/ApoB, Met1206-Asp1413, Accession No. NP_000375 | |
| 100 μg | |
| 338.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry, Immunocytochemistry | |
| 989529 | |
| Immunohistochemistry 5-25 μg;/mL, Immunocytochemistry 8-25 μg;/mL | |
| NP_000375 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Apolipoprotein B/ApoB in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction