Learn More
Human Apolipoprotein B/ApoB Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB41241-SP
 View more versions of this product
Product Code. 18754367
  • € 193.00 / 25µg
Description
Apolipoprotein B/ApoB Monoclonal antibody specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Apolipoprotein B/ApoB
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Apo B-100, APOB, apoB-100, ApoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100
E. coli-derived recombinant human Apolipoprotein B/ApoB, Met1206-Asp1413, Accession No. NP_000375
25 μg
338.0
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG2b
Immunohistochemistry, Immunocytochemistry
989529
Immunohistochemistry 5-25 μg;/mL, Immunocytochemistry 8-25 μg;/mL
NP_000375
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human Apolipoprotein B/ApoB in direct ELISAs.
Human
Supernatant