missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ARHGAP22 (aa 493-560) Control Fragment Recombinant Protein

Produktkod. 30194316
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
Förpackningsstorlek
100µL
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 30194316

missing translation for 'mfr': Invitrogen™ RP103560

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (66%), Rat (66%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64171 (PA5-64171. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

GTPase-activating proteins (GAPs) accelerate the intrinsic rate of GTP hydrolysis of Ras-related proteins, resulting in down regulation of their active form. ARHGAP22 (Rho GTPase activating protein 22), also known as RHOGAP2, is a 698 amino acid protein that localizes to the cytoplasm, and contains one Rho-GAP domain and one PH domain. ARHGAP22 functions as a GTPase-activating protein for Rac 1 converting it to an inactive GDP-bound state and, through its interaction with ZNF161 (VEZF1), is thought to be useful in transcription regulation. Multiple isoforms of ARHGAP22 exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifikationer

Accession Number Q7Z5H3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 58504
Name Human ARHGAP22 (aa 493-560) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Arhgap22; B230341L19Rik; p68RacGAP; RHG22; Rho GTPase activating protein 22; rho GTPase-activating protein 22; RHOGAP2; RhoGap22; Rho-type GTPase-activating protein 22
Common Name ARHGAP22
Gene Symbol Arhgap22
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DNVPAPGLVPGIPSVASMAWSGASSSESSVGGSLSSCTACRASDSSARSSLHTDWALEPSPLPSSSED
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.