missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Arylsulfatase A/ARSA Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF2485
This item is not returnable.
View return policy
Description
Arylsulfatase A/ARSA Polyclonal specifically detects Arylsulfatase A/ARSA in Human samples. It is validated for Western Blot.
Specifications
| Arylsulfatase A/ARSA | |
| Polyclonal | |
| Biotin | |
| AAH14210 | |
| ARSA | |
| Mouse myeloma cell line NS0-derived recombinant human ARSA Arg19-Ala507 Accession # AAH14210 | |
| 50 μg | |
| Primary | |
| Detects human Arylsulfatase A/ARSA in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse ARSA is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| ARSA, arylsulfatase A, ASA, Cerebroside 3-sulfatase, cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 410 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction