missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ASGR2 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB9970N-100UG
This item is not returnable.
View return policy
Description
Asialoglycoprotein Receptor 2 Monoclonal specifically detects Asialoglycoprotein Receptor 2 in Human samples. It is validated for Flow Cytometry.
Specifications
| Asialoglycoprotein Receptor 2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| ASGPR 2, ASGP-R 2, ASGPR2, ASGP-R2, asialoglycoprotein receptor 2, CLEC4H2, CLEC4H2FLJ60040, C-type lectin domain family 4 member H2, HBxAg-binding protein, HBXBP, Hepatic lectin H2, HL-2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ASGR2, Gln80-Ala311, Accession No. P07307 | |
| 100 μg | |
| 433.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 2327C | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| P07307 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human ASGR2 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction